Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MasterTech 95% Reagent Alcohol, StatLab™
SDP

Catalog No. STALREA95GL
Change view
Click to view available options
Quantity:
1 gal./Bottle, 4 Bottles/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STALREA95GL 1 gal./Bottle, 4 Bottles/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STALREA95GL Supplier SLMP Llc Dba Statlab Medical Supplier No. ALREA95GAL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.

95% reagent alcohol without Methyl Alcohol, a known poison that can cause serious health conditions.

  • An effective, safe, alternative option
  • No procedure changes on the user's end
  • Formulated using 100% MasterTech Reagents Alcohol diluted with 5% purified water

Specifications

Physical Form Aqueous Solution
Color Clear
Quantity 1 gal./Bottle, 4 Bottles/Cs.
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.