Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Acetate Buffer ph 8.0
SDP

Catalog No. 22023752
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-752 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-752 Supplier StatLab Supplier No. KC1313
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech Acetate Buffer ph 8.0 - An extraction buffers designed to preserve the integrity of proteins and other biomolecules during extraction processes

  • A buffer designed to preserve the integrity of proteins and other biomolecules during the extraction process
  • A component in the MasterTech Microwave Copper Stain Kit
  • A MasterTech Special Stain Kit Component

Specifications

Chemical Name or Material Acetate Buffer
Grade Laboratory
pH 8
Quantity 1 L
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.