Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Acetate Buffer pH8.0
SDP

Catalog No. 21033381
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-381 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-381 Supplier StatLab Supplier No. KC1312
Add to Cart
Add to Cart

Used to maintain a stable pH during staining procedures

  • Used to maintain a stable pH during staining procedures, ensuring consistent and high-quality staining results by preventing artifacts that could arise from pH fluctuations within the tissue sample
  • Solution is particularly useful in enzyme activity assays, where maintaining the optimal pH for enzyme activity is essential for accurate measurement of reaction rates
  • A component in the MasterTech Microwave Copper Stain Kit
  • A MasterTech Special Stain Kit Component

Specifications

For Use With (Equipment) Staining Equipment
Grade Chemical Solution
Product Type Acetate Buffer
Quantity 16 oz.
Solution Type Buffer
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.