Learn More
StatLab MasterTech Advanced Carbol Fuchsin
SDP
Catalog No. 21033461
Change view
Click to view available options
Volume (Metric):
1000 mL
Catalog No. Volume (Metric)
21-033-461 1000 mL
1 options
Catalog No. 21-033-461 Supplier StatLab Supplier No. STACFLT
Add to Cart
Add to Cart
A 10-minute acid fast bacteria stain.
  • Used to identify Acid Fast Bacteria (mycobacteria), including Mycobacterium tuberculosis and Mycobacterium avium intracellular
  • A MasterTech Special Stain
Specifications
Volume (Metric) 1000 mL
Product Type Advanced Carbol Fuchsin