Learn More
MasterTech Advanced Picric Acid-Acetone - Brown and Brenn, StatLab
SDP
Catalog No. 22035374
Change view
Click to view available options
Quantity:
473 mL
Catalog No. Quantity
22-035-374 473 mL
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-035-374 Supplier SLMP Llc Dba Statlab Medical Supplier No. KC0212
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
A specialized differentiator and counterstain.
  • Its primary function is to differentiate Gram-positive and Gram-negative bacteria within paraffin-embedded tissue sections by providing contrast against the background
  • It removes excess red stain (safranin or basic fuchsin) from the background tissue and Gram-positive bacteria, allowing for better visualization of the bacteria
  • A StatLab MasterTech Special Stain Kit Component
Specifications
Quantity 473 mL
Product Type Picric Acid-Acetone