Learn More
StatLab™ MasterTech AFB Acid Fast Bacteria Stain Kit
SDP
Catalog No. STLKTAFB
Change view
Click to view available options
Quantity:
100 mL
1 pt.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLKTAFB 100 mL
STLKTAFBPT 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLKTAFB Supplier StatLab™ Supplier No. KTAFB
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
AFB Stain Kit provides stains for mycobacteria (acid fast bacteria) in less than 15 minutes.
  • Features Advanced Carbol Fuchsin for bacterial staining
  • Light Green is used as a counterstain
  • 100 mL Stain Kit yields between 80 and 100 slides
  • 1 pt. Stain Kit yields between 320 and 500 slides
Specifications
Quantity 100 mL
Product Type Stain Kit