Learn More
StatLab™ MasterTech AFB Acid Fast Bacteria Stain Kit
SDP
Catalog No. p-200020486
Change view
Click to view available options
Quantity:
100 mL
1 pt.
Catalog No. Quantity
STLKTAFB 100 mL
STLKTAFBPT 1 pt.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLKTAFB
Supplier: StatLab™ KTAFB
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
AFB Stain Kit provides stains for mycobacteria (acid fast bacteria) in less than 15 minutes.
  • Features Advanced Carbol Fuchsin for bacterial staining
  • Light Green is used as a counterstain
  • 100 mL Stain Kit yields between 80 and 100 slides
  • 1 pt. Stain Kit yields between 320 and 500 slides
Specifications
Type Stain Kit
Quantity 100 mL