Learn More
StatLab MasterTech AFB Stain Kit
Description
- This kit is suitable in lieu of and comparable to the Kinyouns AFB and Ziehl-Neelsen procedures for staining all mycobacteria, including Mycobacterium tuberculosis
- The MasterTech AFB Stain Kit differs from these procedures in that Advanced Carbol Fuchsin is used for bacterial staining instead of a standard Carbol Fuchsin, and Light Green is used as a counterstain instead of Methylene Blue
- L Stain Kit yields between 680-1000 slides
- L Kit Consists of: Advanced Carbol Fuchsin L, Light Green Counterstain L, 1% Acid Alcohol 2 L
- A MasterTech Special Stain Kit
Specifications
Specifications
| Product Type | Acid-Fast Bacillus Stain Kit |