Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Albumin, Egg
SDP

Catalog No. 21033470
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-470 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-470 Supplier StatLab Supplier No. TAALB
Add to Cart
Add to Cart

Tissue adhesive for microscope slides, used to stabilize pH, protect cells from stress, and as a substrate for cell attachment

  • Still one of the most popular tissue adhesives used today
  • Just apply a thin coating to the slide
  • Albumin can be used on routine and most special stains
  • It can be used to stabilize pH, protect cells from stress, and as a substrate for cell attachment

Specifications

Physical Form Solution
CAS 56-81-5
Color Colorless to Yellow
Quantity 16 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.