Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Alcian Blue 1.0 pH Stain Kit
SDP

Catalog No. 22021433
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-433 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-433 Supplier StatLab Supplier No. KTABP1
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech Alcian Blue 1.0pH Stain Kit - Stains strongly sulfated mucins blue in 40 minutes

  • This 100 mL stain kit contains a specially formulated Nuclear Fast Red that reduces flocculation not found in similar stain kits on the market that greatly reduces staining artifacts and debris
  • Alcian Blue serves as the primary stain producing vibrant blue stained mucins with the pink to red nuclei counterstained by Nuclear Fast Red
  • There is no filtration required when using MasterTech Nuclear Fast Red
  • 100 mL Stain Kit yields between 80 and 100 slides and consists of - Alcian Blue pH 1.0 100 mL, 3% Acetic Acid Pint, and Nuclear Fast Red Stain 100 mL
  • All kit components should be stored in a dry environment between 15°C and 30°C
  • A MasterTech Special Stain Kit

Specifications

Chemical Name or Material Alcian Blue Stain Kit
Grade Histological
Color Blue
pH 1
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.