Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Alcian Blue Concentrate
SDP

Catalog No. 21033459
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-033-459 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-459 Supplier StatLab Supplier No. STABC25
Add to Cart
Add to Cart

20 drops in 20 ml of water stains weakly sulfated mucosubstances in 30 minutes

  • Used to visualize acidic mucins, which are carbohydrate-containing molecules found in the gastrointestinal and respiratory tracts
  • The dye binds to the acidic mucins, producing a blue to blue-green stain, which is crucial for diagnosing conditions such as Barrett esophagus, mesothelioma, atherosclerosis, and adenocarcinomas
  • A MasterTech Special Stain Kit Component

Specifications

Chemical Name or Material Alcian Blue
CAS 64-17-5, 64-19-7
Grade Laboratory
Color Blue
Quantity 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.