Learn More
StatLab™ MasterTech Alcian Blue Stain
SDP
Catalog No. STSTABL25LT
Change view
Click to view available options
Quantity:
1 pt.
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STSTABL25LT 1 L
STSTABL25PT 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STSTABL25LT Supplier StatLab™ Supplier No. STABL2.5LT
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Alcian Blue stains for weakly sulfated mucosubstances.
  • Used to stain acidic polysaccharides such as glycosaminoglycans in cartilages and other body structures, some types of mucopolysaccharides, sialylated glycocalyx of cells and more
Specifications
Type Alcian Blue
Quantity 1 L