Learn More
StatLab™ MasterTech Alcian Blue Stain
SDP
Catalog No. STSTABL25PT
Change view
Click to view available options
Quantity:
1 pt.
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STSTABL25PT 1 pt.
STSTABL25LT 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STSTABL25PT Supplier StatLab™ Supplier No. STABL2.5PT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Alcian Blue stains for weakly sulfated mucosubstances.
  • Used to stain acidic polysaccharides such as glycosaminoglycans in cartilages and other body structures, some types of mucopolysaccharides, sialylated glycocalyx of cells and more
Specifications
Quantity 1 pt.
Product Type Alcian Blue