Learn More
StatLab™ MasterTech Alcian Blue Stain Kit
SDP
Catalog No. STKTABP25PT
Change view
Click to view available options
Quantity:
100 mL
1 pt.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STKTABP25PT 1 pt.
STLKTABP25 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STKTABP25PT Supplier StatLab™ Supplier No. KTABP2.5PT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Alcian Blue Stain Kit stains acid mucins blue in 40 minutes.
  • With a 2.5 pH, this kit contains a specially formulated Nuclear Fast Red that reduces flocculation, and greatly reduces staining artifacts and debris
  • Alcian Blue serves as the primary stain, producing vibrant blue stained mucins with the pink to red nuclei counterstained by Nuclear Fast Red
  • 100 mL Stain Kit yields between 80 and 100 slides
  • 1 pt. Stain Kit yields between 320 and 500 slides
Specifications
Quantity 1 pt.
Product Type Stain Kit