Learn More
StatLab MasterTech Alcian Blue Stain Kit - pH 1.0

Description
- This kit contains a specially formulated Nuclear Fast Red that reduces flocculation, not found in similar stain kits on the market
- This greatly reduces staining artifacts and debris
- Alcian Blue serves as the primary stain, producing vibrant blue stained mucins with the pink to red nuclei counterstained by Nuclear Fast Red
- There is no filtration required when using MasterTech Nuclear Fast Red
- L Stain Kit yields between 680-1000 slides
- L Kit Consists of: Alcian Blue pH 2.5 L, 3% Acetic Acid two L, and Nuclear Fast Red Stain L
- A MasterTech Special Stain Kit
Specifications
Specifications
| Chemical Name or Material | Alcian Blue Stain Kit |
| CAS | 64-19-7, 64-19-5 |
| Grade | Laboratory |
| Color | Blue/Red |
| pH | 2.5 |
| Quantity | 1000 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.