Learn More
StatLab MasterTech Alcian Blue Stain pH 1.0

Description
- The stain is effective at pH 1.0 for staining sulfated acid mucopolysaccharides and glycoproteins, making it useful for diagnosing conditions such as Barrett’s esophagus, mesothelioma, atherosclerosis, and adenocarcinomas
- It is important to note that Alcian Blue does not stain neutral mucins, and for visualizing neutral mucins, PAS (Periodic Acid Schiff) staining can be used as a counterstain
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| Chemical Name or Material | Ethanol |
| CAS | 64-17-5 |
| Color | Blue |
| pH | Unknown |
| Quantity | 1 Pint |
| Molecular Weight (g/mol) | Unknown |
| Percent Purity | 10% |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.