Learn More
StatLab MasterTech Alcian Blue Stain pH 2.5

Description
- The stain works by binding to the sulfur and/or carboxyl groups of the mucin, depending on the pH used
- With a pH of 2.5, it stains both sulfated and carboxylated acid mucopolysaccharides and glycoproteins, while at a pH of 1.0, it stains only sulfated acid mucopolysaccharides
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| CAS | 64-17-5, 64-19-7 |
| Color | Blue |
| pH | 2.5 |
| Quantity | 100 mL |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.