Learn More
StatLab MasterTech Alcoholic Saffron Solution
SDP
Catalog No. 22021446
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-446 100 mL
22-021-396 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-446 Supplier StatLab Supplier No. KC41A1
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Alcoholic Saffron Solution - used as a special stain to selectively color connective tissue yellow
  • Used as a special stain to selectively color connective tissue yellow particularly in the HPS staining technique
  • A MasterTech Special Stain Kit Component for the MasterTech Russell-Movat Pentachrome Stain Kit
Specifications
Quantity 100 mL
Product Type Alcoholic Saffron Solution