Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Alkaline Congo Red Amyloid Special Stain Kit
SDP

Catalog No. 21010335
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-010-335 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-335 Supplier StatLab Supplier No. KTACR
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Stains for amyloid deposits in less than 15 min.

  • This kit is commonly used to stain for primary, secondary, myeloma-associated, or tumor-associated amyloidosis
  • Alkaline Congo Red Solution serves as the primary stain, producing pink to red amyloid deposits with blue nuclei counterstained by Hematoxylin
  • 100 mL Stain Kit yields between 36 and 45 slides and slide yields assume maximum slides stained per run, maximum runs per use - Actual Slide Yields may vary
  • 100 mL Kit Consists of Alkaline Congo Red Solution pint, Alkaline Alcohol Solution pint, Modified Mayers Hematoxylin 100 mL, Sodium Hydroxide Solution 30 mL
  • All kit components should be stored in a dry environment between 15°C and 30°C
  • Please review all component labels to ensure correct storage temperature
  • StatLab recommends using Amyloid Control Slides
  • A MasterTech Special Stain Kit

Specifications

CAS 1310-73-2, 64-17-5, 573-58-0
Molecular Weight (g/mol) None
Grade Laboratory
Color Red
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.