Learn More
StatLab MasterTech Alkaline Congo Red Solution
SDP
Catalog No. 23640021
Change view
Click to view available options
Quantity:
473 mL
Catalog No. Quantity
23-640-021 473 mL
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-021 Supplier StatLab Supplier No. KC7712
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Used to detect amyloid deposits in tissue sections for histological analysis.
  • Used for the histological detection of amyloid, an extracellular, eosinophilic, proteinaceous material primarily composed of β-pleated sheet structures.
  • Amyloid fibrillar aggregates identified by this stain correspond to structures observed under electron microscopy, supporting accurate pathological correlation.
  • Standard staining process involves deparaffinizing and hydrating tissue sections prior to staining, during which the Congo Red solution binds to amyloid via hydrogen bonding.
  • Provided as a MasterTech Special Stain Kit component, ensuring consistent performance and compatibility within professional histology and pathology laboratories.
Specifications
Quantity 473 mL
Product Type Stain Kit Component