Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Alpha-Amylase
SDP

Catalog No. 21403487
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-403-487 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-403-487 Supplier StatLab Supplier No. SSCAAS250
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Used to determine if glycogen is present in the histology section.

  • This application is essential in differentiating glycogen from other PAS-positive (Periodic Acid-Schiff) substances, such as mucins, fungal walls, or basement membranes
  • A Statlab MasterTech Special Stain Kit component

Specifications

Physical Form Solution
CAS 9000-92-4
Color Colorless
Quantity 250 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.