Learn More
StatLab MasterTech Aqueous Eosin Y, Phloxine B
SDP
Catalog No. 22021386
Change view
Click to view available options
Quantity:
1 gal.
Catalog No. Quantity
22-021-386 1 gal.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-386 Supplier StatLab Supplier No. STE0457
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Aqueous Eosin Y, Phloxine B - A great alternative to traditional alcohol-based formulations
  • The addition of Phloxine B to this aqueous eosin formulation provides a richer warmer H and E counterstain with enhanced differentiation of non-nuclear components
  • Beautiful staining results while eliminating a source of hazard in the lab.
  • Non-Hazardous, Non-Flammable
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 1 gal.
Product Type Eosin Y