Learn More
StatLab MasterTech Biebrich Scarlet/Acid Fuchsin
SDP
Catalog No. 21010252
Change view
Click to view available options
Quantity:
1 gal.
Catalog No. Quantity
21-010-252 1 gal.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-252 Supplier StatLab Supplier No. AHB0457
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Used to differentiate connective tissue components such as collagen and elastic fibers.
  • The dye binds to all tissue components, including muscle and collagen, and is essential for the proper differentiation of these tissues in histological sections
  • The stain is particularly useful in identifying normal tissue morphology and assessing pathologic conditions involving connective tissue
  • A component for the following MasterTech Stain Kits - Massons Trichrome Stain Kit and Massons Trichrome 2000 Stain Kit
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 1 gal.
Product Type Biebrich Scarlet