Learn More
StatLab MasterTech Bielschowsky's Modified Stain Kit
SDP
Catalog No. 21403479
Change view
Click to view available options
Quantity:
6 mL
Catalog No. Quantity
21-403-479 6 mL
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-403-479 Supplier StatLab Supplier No. KT4996
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Used to visualize nerve fibers, axons, neurofibrillary tangles, and senile plaques in brain tissue.
  • Used in research and neuropathology labs to detect, identify, and quantify
  • Alzheimer's disease markers, staining these structures black against a light background
  • A Statlab MasterTech Special Stain Kit
Specifications
Quantity 6 mL
Product Type Stain Kit