Learn More
StatLab MasterTech Bluing Solution

Description
- Solution can be used in a regressive or a progressive H and E staining procedure
- Achieves precise cellular bluing every time
- Used to change the color of hematoxylin-stained nuclei from red to blue
Specifications
Specifications
| For Use With (Equipment) | Staining Equipment |
| Grade | Chemical Solution |
| Product Type | Bluing Reagent |
| Quantity | 1 pt. |
| Solution Type | Stain Kit |
| Form | Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.