Learn More
StatLab MasterTech Brown and Brenn Modified Stain Kit
SDP
Catalog No. 21033388
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
21-033-388 100 mL
1 options
Catalog No. 21-033-388 Supplier StatLab Supplier No. KTBBR
Add to Cart
Add to Cart
Stains for gram positive and gram negative bacteria in just 10 minutes
  • The MasterTech Brown and Brenn Stain Kit is less hazardous than similar kits due to a specially formulated Advanced Picric Acid/Acetone
  • Safranin O and Huckers Gentian Violet both serve as primary stains, producing blue stained gram positive bacteria, along with red gram negative bacteria and nuclei, while the Advanced Picric Acid/Acetone provides a yellow counterstain
  • Pint Stain Kit yields between 144-225 slides
  • Pint Kit Consists of: Gentian Violet pint, 1% Safranin O pint, Universal Iodine Solution pint, ADVANCED Picric Acid/Acetone pint and Gram's Decolorizer pint
  • A MasterTech Special Stain Kit
Specifications
Quantity 100 mL
Product Type Brown and Benn