Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Brown and Brenn Modified Stain Kit
SDP

Catalog No. 22023763
Change view
Click to view available options
Volume (Metric):
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-023-763 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-763 Supplier StatLab Supplier No. KTBBRPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech Brown and Brenn Modified Stain Kit - stains for gram positive and gram negative bacteria in just 10 minutes

  • The MasterTech Brown and Brenn Stain Kit is less hazardous than similar kits due to a specially formulated Advanced Picric Acid/Acetone
  • Safranin O and Hucker's Gentian Violet both serve as primary stains producing blue stained gram positive bacteria along with red gram negative bacteria and nuclei, while the Advanced Picric Acid/Acetone provides a yellow counterstain
  • Pint Stain Kit yields between 144 to 225 slides, actual slide yields may vary
  • Pint Kit Consists of Gentian Violet pint, 1% Safranin O pint, Universal Iodine Solution pint, ADVANCED Picric Acid/Acetone pint, and Grams Decolorizer pint
  • Kit is sealed with a tamper-evident label for quality assurance
  • All kit components should be stored in a dry environment between 15°C and 30°C
  • A MasterTech Special Stain Kit

Specifications

Volume (Metric) 500 mL
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.