Learn More
StatLab™ MasterTech Chandler's Precision Reticulum Stain Kit
SDP
Catalog No. STLKTCPRPT
Change view
Click to view available options
Quantity:
100 mL
1 pt.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLKTCPRPT 1 pt.
STLKTCPR 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLKTCPRPT Supplier StatLab™ Supplier No. KTCPRPT
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Reticulum Stain Kit stains reticulum fibers with no background staining in 30 minutes.
  • Ammoniacal Silver Developer Solution serves as the primary stain, producing brown to black reticulum fibers against a pink to red background counterstained by Nuclear Fast Red
  • No filtration required when using MasterTech Nuclear Fast Red
  • 100 mL Stain Kit yields between 30 and 112 slides
  • 1 pt. Stain Kit yields between 120 and 480 slides
Specifications
Type Stain Kit
Quantity 1 pt.