Learn More
StatLab MasterTech Colloidal Iron Stain Kit
SDP
Catalog No. 21033389
Change view
Click to view available options
Product Type:
Colloidal Iron
Catalog No. Product Type
21-033-389 Colloidal Iron
1 options
Catalog No. 21-033-389 Supplier StatLab Supplier No. KTCIRPT
Add to Cart
Add to Cart
Stains for acid mucosubstances in just 45 minutes
  • Working Colloidal Iron Solution is absorbed and reacts with a working Prussian Blue Solution, serving as the primary stain by producing blue acid mucosubstances against pink to red collagen counterstained by Van Giesons
  • Pint Stain Kit yields between 675-900 slides
  • Pint Kit Consists of: Colloidal Iron Stock 1 pint, Van Gieson's Stain pint, 3% Potassium Ferrocyanide Solution pint, 3% Hydrochloric Acid Solution pint, and 12% Acetic Acid 4 pints
  • A MasterTech Special Stain Kit
Specifications
Product Type Colloidal Iron