Learn More
StatLab MasterTech Colloidal Iron Stock
SDP
Catalog No. 21033380
Change view
Click to view available options
Capacity (Metric):
100 mL
Catalog No. Capacity (Metric)
21-033-380 100 mL
1 options
Catalog No. 21-033-380 Supplier StatLab Supplier No. KC0311
Add to Cart
Add to Cart
Used to identify and visualize acidic mucins within tissue samples
  • Used to identify and visualize acidic mucins within tissue samples, particularly in the lining of the gastrointestinal tract
  • These substances play crucial roles in cellular function, tissue structure, and disease processes
  • The technique involves applying a solution containing positively charged ferric iron (colloidal iron) to bind to the negatively charged groups in acid mucosubstances, followed by a chemical reaction that produces a visible blue precipitate, known as Prussian blue
  • A MasterTech Special Stain Kit Component
Specifications
Capacity (Metric) 100 mL
Product Type Special Stain Kit Component
Autoclavable Not Autoclavable
Disposable Single-use
Graduated No
Mouth Style Rimmed
Sterility Non-sterile
Bottom Shape Round