Learn More
StatLab MasterTech Colloidal Iron Stock
SDP
Catalog No. 22021418
Change view
Click to view available options
Quantity:
1 pt.
Catalog No. Quantity
22-021-418 1 pt.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-418 Supplier StatLab Supplier No. KC0312
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Colloidal Iron Stock a component for the MasterTech Colloidal Iron Stain Kit
  • Used to identify and visualize acidic mucins within tissue samples, particularly in the lining of the gastrointestinal tract
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 1 pt.
Product Type Colloidal Iron Stock Solution