Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Congo Red Amyloid Stain Kit
SDP

Catalog No. 21033391
Change view
Click to view available options
Quantity:
100 mL
16 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-391 16 oz.
21-033-390 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-033-391 Supplier StatLab Supplier No. KTCREPT
Add to Cart
Add to Cart

Stains for amyloid deposits in just over 60 minutes

  • The MasterTech Congo Red Amyloid Kit requires no working solution preparation, making it faster and more convenient than similar kits or procedures
  • Congo Red Amyloid Stain serves as the primary stain, producing pink to red amyloid deposits with blue nuclei counterstained by Hematoxylin
  • 100 mL Stain Kit yields between 320-500 slides
  • 100 mL Kit Consists of: Congo Red Stain 100 mL, Modified Mayer's Hematoxylin 100 mL and 1% Sodium Hydroxide pint
  • A MasterTech Special Stain Kit

Specifications

CAS 64-19-7, 57-55-6, 573-58-0, 1310-73-2
Grade Laboratory
Color Pink to Red
Quantity 16 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.