Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Copper Stain Kit, Microwave
SDP

Catalog No. 21033401
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-401 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-401 Supplier StatLab Supplier No. KTMCO
Add to Cart
Add to Cart

Stains for copper in just 10 minutes with 100 ml stain kit yields between 680 and 1000 slides

  • This kit includes a 10 minute microwave procedure and the traditional 18 hour procedure
  • Rhodanine Stock Solution serves as the primary stain, producing light brown to red cytoplasmic copper granules with blue nuclei counterstained by Hematoxylin
  • 100 mL Stain Kit yields between 680 and 1000 slides
  • 100 mL Kit Consists of: Rhodanine Stock Stain Solution 25ml, Acetate Buffer pH 8.0 2 Pints, and Modified Mayer's Hematoxylin 100 mL
  • A MasterTech Special Stain Kit

Specifications

Quantity 100 mL
Product Type Copper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.