Learn More
StatLab MasterTech Copper Stain Kit

Description
- This kit includes a 10 minute microwave procedure and the traditional 18 hour procedure
- Rhodanine Stock Solution serves as the primary stain producing light brown to red cytoplasmic copper granules with blue nuclei counterstained by Hematoxylin
- Pint Stain Kit yields between 1500-2000 slides and actual slide yields may vary
- Pint Kit Consists of Rhodanine Stock Stain Solution 100 mL, Acetate Buffer pH8 0.4 pint, and Modified Mayers Hematoxylin pint
- Kit is sealed with a tamper-evident label for quality assurance and all kit components should be stored in a dry environment between 15°C and 30°C
- Please review all component labels to ensure correct storage temperature
- A MasterTech Special Stain Kit
Specifications
Specifications
| Color | Brown to Red |
| Quantity | 1 pt. |
| Grade | Histological |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.