Learn More
StatLab MasterTech Cresyl Echt Violet Solution
SDP
Catalog No. 23640008
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
23-640-008 100 mL
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-008 Supplier StatLab Supplier No. AHC0426
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Special stain solution used to visualize Nissl substance in neurons, supporting assessment of neuronal structure and integrity.
  • Cresyl Echt Violet Solution formulated to stain Nissl substance, which is composed of rough endoplasmic reticulum and ribosomes, serving as key markers for evaluating neuronal structure and cellular integrity in histological sections.
  • Commonly used in neuropathology applications, this solution also supports the evaluation and assessment of neurodegeneration in nervous tissue samples.
  • Validated component used within MasterTech stain kits, including the Pneumocystis Stain Kit CN1.5 and the Luxol Fast Blue-Cresyl Echt Violet Stain Kit, ensuring compatibility and reliable staining performance.
  • Supplied as a MasterTech Special Stain Kit component, suitable for professional histology laboratories requiring consistent and reproducible neuronal staining results.
Specifications
Quantity 100 mL
Product Type Stain Kit Component