Learn More
StatLab MasterTech Cresyl Echt Violet Stain

Description
- Used to stain the cell bodies and processes of neurons, specifically highlighting the Nissl substance (rough endoplasmic reticulum) within them, which allows for the identification and visualization of neuronal structure in tissue sections
- A component used for the following MasterTech Stain Kits: Pneumocystis Stain Kit CN1.5 and Luxol Fast Blue-Cresyl Echt Violet Stain Kit
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| CAS | 64-17-5 |
| Color | Dark Blue |
| Quantity | 16 oz. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.