Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Cresyl Echt Violet
SDP

Catalog No. 22021441
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-441 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-441 Supplier StatLab Supplier No. AHCEV25
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Cresyl Echt Violet, a MasterTech Special Stain Kit Comonent

  • Used to stain the cell bodies and processes of neurons, specifically highlighting the Nissl substance (rough endoplasmic reticulum) within them which allows for the identification and visualization of neuronal structure in tissue sections
  • Compoent is often used in conjunction with other stains like Luxol Fast Blue to visualize both neurons and myelin sheaths in nervous tissue
  • A MasterTech Special Stain Kit Component

Specifications

Grade Laboratory
Color Violet
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.