Learn More
StatLab MasterTech Crocein Scarlet Acid Fuchsin
SDP
Catalog No. 22021366
Change view
Click to view available options
Quantity:
1 pt.
Catalog No. Quantity
22-021-366 1 pt.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-366 Supplier StatLab Supplier No. KC41B2
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Crocein Scarlet Acid Fuchsin - used to stain connective tissue fibers like collagen
  • Used to stain connective tissue fibers like collagen particularly in procedures like Massons Trichrome stain
  • A MasterTech Special Stain Kit Component for the MasterTech Russell-Movat Pentachrome Stain Kit
Specifications
Quantity 1 pt.
Product Type Crocein Scarlet Acid Fuchsin