Learn More
StatLab MasterTech Crystal Violet Amyloid Stain
SDP
Catalog No. 22024800
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
22-024-800 100 mL
1 options
Catalog No. 22-024-800 Supplier StatLab Supplier No. STCVA100
Add to Cart
Add to Cart
Designed to rapidly screen for amyloid deposits
  • Stains amyloid deposits a distinct purple-violet color while staining surrounding tissues blue
  • It is frequently used for detecting amyloid in tissue samples as an alternative to Congo red, though it is less specific and sometimes used for preliminary screening
  • A StatLab MasterTech Special Stain Kit Component
Specifications
Quantity 100 mL