Learn More
StatLab MasterTech Differentiating Solution
Description
- Allows for a clearer distinction between different tissue components by ensuring only the desired structures retain the stain
- Essentially fine-tunes the staining process - this is particularly important when using regressive staining techniques where the tissue is initially overstained
Specifications
Specifications
| Product Type | Differentiating Solution |