Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Easycut Nail Softening Solution
SDP

Catalog No. 21033471
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-471 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-471 Supplier StatLab Supplier No. TCZ00975E
Add to Cart
Add to Cart

Fast-acting Formulation for nail softening, fixed tissue that is overhardened and difficult to section

  • Use EasyCut Softening Solution on keratin, blood, muscle, skin, decalcified bone, or any fixed tissue that is overhardened and difficult to section
  • Just soak tissue blocks for a few seconds and begin sectioning

Specifications

Physical Form Solution
CAS 7647-01-0
Color Clear Colorless
Quantity 100 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.