Learn More
StatLab MasterTech Eosin Stain
SDP
Catalog No. 22021427
Change view
Click to view available options
Quantity:
1 gal.
Catalog No. Quantity
22-021-427 1 gal.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-427 Supplier StatLab Supplier No. STE0157
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Eosin Stain - Stains cytoplasmic structures, collagen, and basement membranes with three distinct shades of pink
  • Eosin Y is used to stain eosinophilic compounds, which are positively charged and contain eosinophilic compounds
  • It is commonly used in conjunction with hematoxylin to demonstrate the general histological architecture of tissues
  • Eosin Y is soluble in water and is particularly useful for staining cytoplasm, red blood cells, collagen, and muscle fibers
  • The dye's ability to distinguish between different cell types and structures in shades of red and pink makes it an essential tool in histopathology for visualizing tissue structures and identifying various histopathologic disorders
  • A MasterTech Stain
Specifications
Quantity 1 gal.
Product Type Eosin Stain