Learn More
StatLab MasterTech Eosin Y Alcoholic Stain

Description
- Ready-to-use formulation for routine HandE Staining
- Stain aids in the visualization of erythrocytes, collagen, and the cytoplasm of muscle or epithelial cells
- The use of Eosin Y in histology is essential for pathologists to distinguish between different tissue components, making it a cornerstone in disease diagnosis, especially cancer
- A MasterTech HandE Stain
Specifications
Specifications
| Chemical Name or Material | Eosyin Y Stain |
| CAS | 64-17-5, 17372-87-1 |
| Color | Red |
| Quantity | 16 oz. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.