Learn More
StatLab MasterTech Eosin Y Stain
SDP
Catalog No. 22000082
Change view
Click to view available options
Quantity:
Gallon
Pint
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-000-082 Gallon
22-000-081 Pint
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-082 Supplier StatLab Supplier No. STE0257
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
The addition of Phloxine B to this MasterTech Alcoholic Eosin formulation provides a richer warmer H and E counterstain with enhanced differentiation of non-nuclear components
  • The addition of Phloxine B to this MasterTech Alcoholic Eosin formulation provides a richer warmer H and E counterstain
  • A MasterTech Eosin Stain
  • Ready-to-use
Specifications
Quantity Gallon
Product Type Eosin Y Stain Phloxine B