Learn More
StatLab MasterTech Eosin Y Stain
SDP
Catalog No. 23640022
Change view
Click to view available options
Quantity:
5 gal.
Catalog No. Quantity
23-640-022 5 gal.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-022 Supplier StatLab Supplier No. STE0159
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Primarily used to provide clear contrast between acidophilic cytoplasmic structures and basophilic nuclear structures.
  • Eosin Y stain formulated to provide clear contrast between basic (acidophilic) structures of the cytoplasm and acidic (basophilic) structures of the nuclei, supporting detailed visualization of cells and tissues.
  • This contrasting effect is essential for accurate assessment of cellular detail and overall tissue architecture, aiding in disease diagnosis and the evaluation of tissue changes.
  • Particularly significant in the diagnosis of certain cancers and other pathological conditions where precise differentiation between normal and abnormal tissue is critical.
  • Supplied as a MasterTech Special Stain Kit component, ensuring consistent quality, compatibility, and reliable performance in histology laboratories.
Specifications
Quantity 5 gal.
Product Type Stain Kit Component