Learn More
StatLab MasterTech Eosin Y Stain, Aqueous
SDP
Catalog No. 22046657
Change view
Click to view available options
Packaging:
Bottle, 1 gal.
Catalog No. Packaging
22-046-657 Bottle, 1 gal.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-046-657 Supplier StatLab Supplier No. STE0357
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Non-hazardous and non-flammable alternative to traditional alcohol-based Eosin-Y formulations.
  • Non-hazardous and non-flammable alternative to traditional alcohol-based Eosin-Y formulations.
  • Produces beautiful cytoplasmic staining while eliminating a source of hazard in the lab.
  • Ready-to-use.
  • Available in 1-gallon StatGrip, rectangular F-style bottles.
  • HistoFlow dispensing technology designed for easy pouring, improved ergonomics, and smarter storage.
Specifications
Color Clear
Packaging Bottle, 1 gal.
Quantity 1 gal.