Learn More
StatLab MasterTech Eosin Y Stain, Phloxine B

Description
- The addition of Phloxine B to this MasterTech Alcoholic Eosin formulation provides a richer warmer HandE counterstain with enhanced differentiation of non-nuclear components
- Ready-to-use formulation also works well for cytoplasmic staining
- A MasterTech HandE Stain
Specifications
Specifications
| Chemical Name or Material | Eosin Y Phoxine B |
| CAS | 64-17-5 |
| Color | Red |
| Quantity | 1000 mL |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.