Learn More
StatLab MasterTech Feulgen Stain Kit
SDP
Catalog No. 21033392
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
21-033-392 100 mL
1 options
Catalog No. 21-033-392 Supplier StatLab Supplier No. KTFEU
Add to Cart
Add to Cart
Stains for deoxyribonucleic acid (DNA) in 35 minutes
  • Also known as the Feulgen Reaction, hydrochloric acid removes the purine bases from the DNA, leaving the sugars and phosphates
  • This creates an aldehyde group that can be demonstrated by the Optimized Schiffs Solution
  • The MasterTech Feulgen Stain kit is up to two times faster than similar procedures, sometimes requiring 60 minutes alone in the Schiffs
  • Optimized Schiffs Solution serves as the primary stain, producing red to purple stained DNA against a light green background counterstained by Fast Green
  • 100 mL Stain Kit yields between 144 and 225 slides
  • 100 mL Kit Consists of: 1.0N Hydrochloric Acid Pint, Sulfurous Rinse 2 Pints, Optimized Schiffs Solution Pint, 0.2% Fast Green 100 mL
  • A MasterTech Special Stain Kit
Specifications
Color Assorted
Quantity 100 mL
Product Type Special Stain Kit