Learn More
StatLab MasterTech Fite's Stain Kit, Modified - Leprosy Stain
SDP
Catalog No. 21033394
Change view
Click to view available options
Quantity:
100 mL
16 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-394 16 oz.
21-033-393 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-033-394 Supplier StatLab Supplier No. KTFITPT
Add to Cart
Add to Cart
Stains for mycobacterium leprae and nocardia in less than 30 minutes
  • This kit is commonly used demonstrate the causative organism of leprosy and differs from similar kits or procedures in that Advanced Carbol Fuchsin (ACF) is used instead of a standard Carbol Fuchsin and Light Green is used as a counterstain instead of Methylene Blue
  • In comparison to standard Carbol Fuchsins, MasterTech ACF requires no heat and has a much shorter staining time by combining all three fuchsin dyes
  • Light Green is a much better counterstain than Methylene Blue, as it provides higher contrast against the red stained bacteria
  • Advanced Carbol Fuchsin serves as the primary stain, producing red bacteria against a blue background counterstained by Light Green
  • 100 mL Stain Kit yields between 53 and 66 slides
  • 100 mL Kit Consists of: Xylene Peanut Oil 100 mL, Advanced Carbol Fuchsin 100 mL, Light Green Counterstain 100 mL and 1% Acid Alcohol Pint
  • A MasterTech Special Stain Kit
Specifications
Quantity 16 oz.
Product Type Fite's Method Stain Kit