Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Formalin 20% Reticulum
SDP

Catalog No. 21279191
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-279-191 100 mL
21-279-192 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-279-191 Supplier StatLab Supplier No. KC3941
Add to Cart
Add to Cart
This item is not returnable. View return policy

Stain Kit Component, for tissue preservation

  • Used for tissue preservation
  • Stabilizes proteins
  • Prevents autolysis and putrefaction
  • A common fixative for general histology
  • A StatLab MasterTech Stain Kit Component

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Formalin
Quantity 100 mL
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.